2,3,4-Trifluorobenzonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
PC7283D-100G
PC7283D-25G
PC7283D-5G
APOSPC7283D-100GEA
68.3
EUR
APOSPC7283D-100G
APOSPC7283D-25G
APOSPC7283D-5G
2,3,4-Trifluorobenzonitrile 98%
2,3,4-Trifluorobenzonitrile
Formula:
C₇H₂F₃N Boiling Pt: 116…118 °C/100mmHg Density: 1,12 g/cm³ (25 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013288 CAS Number: 143879-80-5 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...