α,α,α,3,4,5-Hexafluorotoluene 97%

Supplier: Apollo Scientific
Warning

PC7284T-25G PC7284T-5G PC7284T-10G
APOSPC7284T-25GEA 375 EUR
APOSPC7284T-25G APOSPC7284T-5G APOSPC7284T-10G
α,α,α,3,4,5-Hexafluorotoluene 97%
α,α,α,3,4,5-Hexafluorotoluene
Formula: C₇H₂F₆
MW: 200,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153280
CAS Number: 80172-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR