2,4,5-Trifluorobenzyl alcohol 97%

Supplier: Apollo Scientific
Warning

PC7285LD-25G PC7285LD-5G
APOSPC7285LD-25GEA 72.5 EUR
APOSPC7285LD-25G APOSPC7285LD-5G
2,4,5-Trifluorobenzyl alcohol 97%
2,4,5-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
Boiling Pt: 201…204 °C
Density: 1,4 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061206
CAS Number: 144284-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR