4,4,4-Trifluoro-2-butanone 97%

Supplier: Apollo Scientific
Warning

PC7290-1G PC7290-5G
APOSPC7290-1GEA 66.7 EUR
APOSPC7290-1G APOSPC7290-5G
4,4,4-Trifluoro-2-butanone 97%
4,4,4-Trifluoro-2-butanone
Formula: C₄H₅F₃O
MW: 126,08 g/mol
Boiling Pt: 95…96 °C
Density: 1,05 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00077601
CAS Number: 2366-70-3
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR