2-Furoyltrifluoroacetone 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4,4,4-Trifluoro-1-(furan-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione
PC7380-5G
PC7380-25G
APOSPC7380-5GEA
14.7
EUR
APOSPC7380-5G
APOSPC7380-25G
2-Furoyltrifluoroacetone 99%
2-Furoyltrifluoroacetone
Formula:
C₈H₅F₃O₃ MW: 206,12 g/mol Boiling Pt: 203 °C Melting Pt: 20…21 °C Density: 1,391 g/cm³ (25 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020935 CAS Number: 326-90-9 EINECS: 206-315-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...