4-(Trifluoromethoxy)benzaldoxime 97%

Supplier: Apollo Scientific
Warning

PC7434L-1G PC7434L-250MG
APOSPC7434L-1GEA 109 EUR
APOSPC7434L-1G APOSPC7434L-250MG
4-(Trifluoromethoxy)benzaldoxime 97%
4-(Trifluoromethoxy)benzaldoxime
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00210176
CAS Number: 150162-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR