4-(Trifluoromethoxy)phenylacetonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(Cyanmethyl)-4-(trifluormethoxy)benzol, 4-(TrifluoroMethoxy)phenylacetonitrile, 4-(Trifluoromethoxy)benzyl cyanide
PC7440H-100G
PC7440H-25G
PC7440H-5G
APOSPC7440H-100GEA
128
EUR
APOSPC7440H-100G
APOSPC7440H-25G
APOSPC7440H-5G
4-(Trifluoromethoxy)phenylacetonitrile 98%
4-(Trifluoromethoxy)phenylacetonitrile
Formula:
C₉H₆F₃NO Boiling Pt: 96 °C/3,75mmHg Melting Pt: 28…33 °C Density: 1,28 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061240 CAS Number: 49561-96-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...