3'-(Trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
PC7480-100G
PC7480-25G
PC7480-500G
APOSPC7480-100GEA
59.6
EUR
APOSPC7480-100G
APOSPC7480-25G
APOSPC7480-500G
3'-(Trifluoromethyl)acetophenone 98%
3'-(Trifluoromethyl)acetophenone
Formula:
C₉H₇F₃O MW: 188,15 g/mol Boiling Pt: 198…200 °C Density: 1,235 g/cm³ (25 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000391 CAS Number: 349-76-8 EINECS: 206-490-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...