4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromomethyl-3-fluorophenylboronic acid pinacol ester, 2-(4-(Bromomethyl)-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-Dioxaborolane, 2-[4-(bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-, 2-[4-(Bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PC7518-1G
PC7518-250MG
PC7518-5G
APOSPC7518-1GEA
219
EUR
APOSPC7518-1G
APOSPC7518-250MG
APOSPC7518-5G
4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester 95+%
4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester
Formula:
C₁₃H₁₇BBrFO₂ MW: 314,99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11855987 CAS Number: 1150271-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...