2-Bromo-4-nitro-6-(trifluoromethyl)aniline 95+%

Supplier: Apollo Scientific

Synonyms: 2-Amino-3-bromo-5-nitrobenzotrifluoride

PC7524-1G PC7524-500MG PC7524-5G
APOSPC7524-1GEA 212 EUR
APOSPC7524-1G APOSPC7524-500MG APOSPC7524-5G
2-Bromo-4-nitro-6-(trifluoromethyl)aniline 95+%
2-Bromo-4-nitro-6-(trifluoromethyl)aniline
Formula: C₇H₄BrF₃N₂O₂
MW: 285.0200 g/mol
Melting Pt: 142…143 °C
Storage Temperature: Ambient
MDL Number: MFCD14560537
CAS Number: 400-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR