1-[(2-Cyano-4-fluoro)phenyl]piperidine-3-carboxylic acid

Supplier: Apollo Scientific

PC7559-1G PC7559-5G
APOSPC7559-1GEA 126 EUR
APOSPC7559-1G APOSPC7559-5G
1-[(2-Cyano-4-fluoro)phenyl]piperidine-3-carboxylic acid
1-[(2-Cyano-4-fluoro)phenyl]piperidine-3-carboxylic acid
Formula: C₁₃H₁₃FN₂O₂
MW: 248,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750917
CAS Number: 1000339-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR