3-Fluoro-4-nitrobenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC7624-1G
PC7624-250MG
PC7624-5G
APOSPC7624-1GEA
39.3
EUR
APOSPC7624-1G
APOSPC7624-250MG
APOSPC7624-5G
3-Fluoro-4-nitrobenzaldehyde 98%
3-Fluoro-4-nitrobenzaldehyde
Formula:
C₇H₄NO₃F MW: 169.1100 g/mol Melting Pt: 57…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00968940 CAS Number: 160538-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...