2-(Trifluoromethyl)benzylamine

Supplier: Apollo Scientific
Danger

Synonyms: (2-(Trifluoromethyl)phenyl)methanamine

PC7628-100G PC7628-25G PC7628-5G
APOSPC7628-100GEA 259 EUR
APOSPC7628-100G APOSPC7628-25G APOSPC7628-5G
2-(Trifluoromethyl)benzylamine
2-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175,15 g/mol
Boiling Pt: 108…110 °C/64mmHg
Density: 1,249 g/cm³ (25 °C)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00010297
CAS Number: 3048-01-9
EINECS: 221-258-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR