4-Bromo-5-fluoro-2-nitrotoluene 99%

Supplier: Apollo Scientific
Warning

PC7643-25G PC7643-5G PC7643-100G
APOSPC7643-25GEA 46.6 EUR
APOSPC7643-25G APOSPC7643-5G APOSPC7643-100G
4-Bromo-5-fluoro-2-nitrotoluene 99%
4-Bromo-5-fluoro-2-nitrotoluene
Formula: C₇H₅BrFNO₂
MW: 234,02 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD04039299
CAS Number: 224185-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR