(α,α,α-Trifluoro-2-tolyl)acetic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Trifluoromethyl)phenylacetic acid, (2-(Trifluoromethyl)phenyl)acetic acid

PC7698-5G PC7698-1G PC7698-10G
APOSPC7698-5GEA 21.9 EUR
APOSPC7698-5G APOSPC7698-1G APOSPC7698-10G
(α,α,α-Trifluoro-2-tolyl)acetic acid 98%
(α,α,α-Trifluoro-2-tolyl)acetic acid
Formula: F₃CC₆H₄CH₂COOH
MW: 204,15 g/mol
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00004327
CAS Number: 3038-48-0
EINECS: 221-240-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR