2-Fluorobenzylamine 95%
Supplier: Apollo Scientific
|
Danger
|
PC7734-100G
PC7734-25G
PC7734-500G
APOSPC7734-100GEA
201
EUR
APOSPC7734-100G
APOSPC7734-25G
APOSPC7734-500G
2-Fluorobenzylamine 95%
2-Fluorobenzylamine
Formula:
C₇H₈FN MW: 125,15 g/mol Boiling Pt: 73…75 °C/13mmHg Density: 1,099 g/cm³ (25 °C) Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008107 CAS Number: 89-99-6 EINECS: 201-957-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...