Pentafluoropropylamidine 95%

Supplier: Apollo Scientific
Warning

Synonyms: Perfluoropropanamidine

PC7737-25G PC7737-5G PC7737-1G PC7737-10G
APOSPC7737-25GEA 0 EUR
APOSPC7737-25G APOSPC7737-5G APOSPC7737-1G APOSPC7737-10G
Pentafluoropropylamidine 95%
Pentafluoropropylamidine
Formula: C₃H₃F₅N₂
MW: 162.06 g/mol
Melting Pt: 49…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06248648
CAS Number: 422-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR