2-Hydrazinyl-5-(trifluoromethyl)pyridine 97%

Supplier: Apollo Scientific
Warning

PC7766X-1G PC7766X-250MG
APOSPC7766X-1GEA 24.8 EUR
APOSPC7766X-1G APOSPC7766X-250MG
2-Hydrazinyl-5-(trifluoromethyl)pyridine 97%
2-Hydrazinyl-5-(trifluoromethyl)pyridine
Formula: C₆H₆F₃N₃
Storage Temperature: Refrigerator
MDL Number: MFCD00236724
CAS Number: 89570-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR