(2,3,6-Trifluorophenyl)acetic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(2,3,6-Trifluorophenyl)acetic acid
PC7818E-5G
PC7818E-1G
APOSPC7818E-5GEA
48
EUR
APOSPC7818E-5G
APOSPC7818E-1G
(2,3,6-Trifluorophenyl)acetic acid 97%
(2,3,6-Trifluorophenyl)acetic acid
Formula:
C₆H₂F₃CH₂COOH MW: 190,12 g/mol Melting Pt: 111…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061217 CAS Number: 114152-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...