(3,4,5-Trifluorophenyl)acetic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,4,5-Trifluorophenylacetic acid
PC7818H-1G
PC7818H-5G
PC7818H-10G
APOSPC7818H-1GEA
21.9
EUR
APOSPC7818H-1G
APOSPC7818H-5G
APOSPC7818H-10G
(3,4,5-Trifluorophenyl)acetic acid 97%
(3,4,5-Trifluorophenyl)acetic acid
Formula:
(C₆H₂F₃)CH₂COOH MW: 190,12 g/mol Melting Pt: 98…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00083532 CAS Number: 209991-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...