N-(2,3,4-Trifluorophenyl)thiourea 97%
Supplier: Apollo Scientific
|
Warning
|
PC7819H-10G
PC7819H-1G
APOSPC7819H-10GEA
176
EUR
APOSPC7819H-10G
APOSPC7819H-1G
N-(2,3,4-Trifluorophenyl)thiourea 97%
N-(2,3,4-Trifluorophenyl)thiourea
Formula:
C₇H₅F₃N₂S MW: 206,19 g/mol Boiling Pt: 162…165 °C Melting Pt: 162…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053006 CAS Number: 175205-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...