1,1,1-Trifluoropropane 97%

Supplier: Apollo Scientific
Warning

PC7819M-25G PC7819M-5G PC7819M-10G
APOSPC7819M-25GEA 630 EUR
APOSPC7819M-25G APOSPC7819M-5G APOSPC7819M-10G
1,1,1-Trifluoropropane 97%
1,1,1-Trifluoropropane
Formula: C₃H₅F₃
Boiling Pt: –12 °C
Melting Pt: –148 °C
Storage Temperature: Ambient
CAS Number: 421-07-8
UN: 3161
ADR: 2.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR