2-Fluoro-6-(methylamino)benzenecarbonitrile 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Fluoro-6-(methylamino)benzonitrile
PC7924-1G
APOSPC7924-1GEA
0
EUR
APOSPC7924-1G
2-Fluoro-6-(methylamino)benzenecarbonitrile 95%
2-Fluoro-6-(methylamino)benzenecarbonitrile
Formula:
C₈H₇FN₂ MW: 150,16 g/mol Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD12193344 CAS Number: 96783-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...