2-Fluoro-3-methoxybenzyl bromide
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(Bromomethyl)-2-fluoro-3-methoxybenzene
PC7985-1G
PC7985-25G
PC7985-5G
APOSPC7985-1GEA
55.1
EUR
APOSPC7985-1G
APOSPC7985-25G
APOSPC7985-5G
2-Fluoro-3-methoxybenzyl bromide
2-Fluoro-3-methoxybenzyl bromide
Formula:
C₈H₈BrFO Boiling Pt: 126…127 °C/10mmHg Melting Pt: 45…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD09743597 CAS Number: 447463-56-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...