5-Fluoro-3-iodo-1-methyl-1H-indazole
Supplier: Apollo Scientific
|
Warning
|
PC7999-1G
PC7999-5G
APOSPC7999-1GEA
203
EUR
APOSPC7999-1G
APOSPC7999-5G
5-Fluoro-3-iodo-1-methyl-1H-indazole
5-Fluoro-3-iodo-1-methyl-1H-indazole
Formula:
C₈H₆FIN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11053924 CAS Number: 1060817-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...