Zinc trifluoromethanesulphonate 98%

Supplier: Apollo Scientific
Danger

Synonyms: Zinc(II) trifluoromethanesulphonate, Zinc triflate, Zinc trifluoromethanesulphonate

PC8034-100G PC8034-25G PC8034-5G
APOSPC8034-100GEA 152 EUR
APOSPC8034-100G APOSPC8034-25G APOSPC8034-5G
Zinc trifluoromethanesulphonate 98%
Zinc trifluoromethanesulphonate
Formula: C₂F₆O₆S₂Zn
MW: 363,53 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013229
CAS Number: 54010-75-2
EINECS: 258-922-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR