4-Amino-8-(trifluoromethyl)quinoline

Supplier: Apollo Scientific
Warning

Synonyms: 8-(Trifluoromethyl)quinolin-4-amine

PC8045-1G PC8045-250MG
APOSPC8045-1GEA 112 EUR
APOSPC8045-1G APOSPC8045-250MG
4-Amino-8-(trifluoromethyl)quinoline
4-Amino-8-(trifluoromethyl)quinoline
Formula: C₁₀H₇F₃N₂
MW: 212,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269346
CAS Number: 243977-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR