7-(Trifluoromethyl)-1H-indazole

Supplier: Apollo Scientific
Warning

PC8057-1G PC8057-5G PC8057-250MG
APOSPC8057-1GEA 105 EUR
APOSPC8057-1G APOSPC8057-5G APOSPC8057-250MG
7-(Trifluoromethyl)-1H-indazole
7-(Trifluoromethyl)-1H-indazole
Formula: C₈H₅F₃N₂
Melting Pt: 92…99 °C
Storage Temperature: Ambient
MDL Number: MFCD09836171
CAS Number: 885694-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR