2-(Methylthio)-5-(trifluoromethoxy)phenylboronic acid

Supplier: Apollo Scientific

PC8077-1G PC8077-250MG
APOSPC8077-1GEA 288 EUR
APOSPC8077-1G APOSPC8077-250MG
2-(Methylthio)-5-(trifluoromethoxy)phenylboronic acid
2-(Methylthio)-5-(trifluoromethoxy)phenylboronic acid
Formula: C₈H₈BF₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD09800888
CAS Number: 957121-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR