4-Fluoro-2-(trifluoromethyl)cinnamic acid 97%

Supplier: Apollo Scientific
Warning

PC8123-1G PC8123-10G PC8123-5G
APOSPC8123-1GEA 29 EUR
APOSPC8123-1G APOSPC8123-10G APOSPC8123-5G
4-Fluoro-2-(trifluoromethyl)cinnamic acid 97%
4-Fluoro-2-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆F₄O₂
MW: 234,15 g/mol
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00236297
CAS Number: 243977-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR