2-Chloro-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide
Supplier: Apollo Scientific
|
Warning
|
PC8133-1G
PC8133-500MG
PC8133-5G
APOSPC8133-1GEA
533
EUR
APOSPC8133-1G
APOSPC8133-500MG
APOSPC8133-5G
2-Chloro-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide
2-Chloro-N-(2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)acetamide
Formula:
C₁₀H₁₀Cl₂F₃N₃O Melting Pt: 105…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126811 CAS Number: 339096-66-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...