N-Formyl 2-chloro-5-(trifluoromethyl)aniline

Supplier: Apollo Scientific

Synonyms: N-Formyl 2-chloro-5-(trifluoromethyl)aniline, 6'-CHLORO-ALPHA,ALPHA,ALPHA-TRIFLUORO-M-FORMOTOLUIDIDE, N-(2-Chloro-5-(trifluoromethyl)phenyl)formamide

PC8143-1G PC8143-5G
APOSPC8143-1GEA 118 EUR
APOSPC8143-1G APOSPC8143-5G
N-Formyl 2-chloro-5-(trifluoromethyl)aniline
N-Formyl 2-chloro-5-(trifluoromethyl)aniline
Formula: C₈H₅ClF₃NO
MW: 223,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021000
CAS Number: 657-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR