1-Chloro-4-fluoro-3-iodobenzene 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Chloro-1-fluoro-2-iodobenzene
PC8156-25G
PC8156-5G
APOSPC8156-25GEA
56.7
EUR
APOSPC8156-25G
APOSPC8156-5G
1-Chloro-4-fluoro-3-iodobenzene 97%
1-Chloro-4-fluoro-3-iodobenzene
Formula:
C₆H₃ClFI MW: 256,4400 g/mol Boiling Pt: 214 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00672963 CAS Number: 116272-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...