4,5-Dibromo-2-fluorotoluene

Supplier: Apollo Scientific
Warning

PC8178-25G PC8178-5G
APOSPC8178-25GEA 116 EUR
APOSPC8178-25G APOSPC8178-5G
4,5-Dibromo-2-fluorotoluene
4,5-Dibromo-2-fluorotoluene
Formula: C₇H₅Br₂F
MW: 267,92 g/mol
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00143234
CAS Number: 202982-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR