4-Bromo-1-(4-fluorophenyl)-1H-pyrazole
Supplier: Apollo Scientific
|
Warning
|
PC8337-1G
PC8337-250MG
PC8337-5G
APOSPC8337-1GEA
120
EUR
APOSPC8337-1G
APOSPC8337-250MG
APOSPC8337-5G
4-Bromo-1-(4-fluorophenyl)-1H-pyrazole
4-Bromo-1-(4-fluorophenyl)-1H-pyrazole
Formula:
C₉H₆BrFN₂ MW: 241,0600 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09801028 CAS Number: 957062-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...