2-Chloro-1,1,1,4,4,4-hexafluorobutane 97%

Supplier: Apollo Scientific

PC8342-25G PC8342-5G PC8342-1G
APOSPC8342-25GEA 889 EUR
APOSPC8342-25G APOSPC8342-5G APOSPC8342-1G
2-Chloro-1,1,1,4,4,4-hexafluorobutane 97%
2-Chloro-1,1,1,4,4,4-hexafluorobutane
Formula: C₄H₃ClF₆
Boiling Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD01320788
CAS Number: 400-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR