5-Fluoro-3-nitropyridin-2(1H)-one

Supplier: Apollo Scientific
Warning

Synonyms: 5-Fluoro-2-hydroxy-3-nitropyridine

PC8393-1G PC8393-5G
APOSPC8393-1GEA 21.9 EUR
APOSPC8393-1G APOSPC8393-5G
5-Fluoro-3-nitropyridin-2(1H)-one
5-Fluoro-3-nitropyridin-2(1H)-one
Formula: C5H3FN2O3
MW: 158.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662412
CAS Number: 136888-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR