2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-phenylacetonitrile 97%

Supplier: Apollo Scientific

PC8419-1G PC8419-500MG PC8419-5G
APOSPC8419-1GEA 275 EUR
APOSPC8419-1G APOSPC8419-500MG APOSPC8419-5G
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-phenylacetonitrile 97%
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-phenylacetonitrile
Formula: C₁₄H₈ClF₃N₂
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00172206
CAS Number: 213994-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR