2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}nicotinic acid 97%
Supplier: Apollo Scientific
|
Warning
|
PC8434-1G
PC8434-5G
APOSPC8434-1GEA
87
EUR
APOSPC8434-1G
APOSPC8434-5G
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}nicotinic acid 97%
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}nicotinic acid
Formula:
C₁₂H₆ClF₃N₂O₂S MW: 334,71 g/mol Melting Pt: 183…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00244738 CAS Number: 303149-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...