2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(2-Cyano-3-fluoro)-2-(thien-2-yl)acetonitrile
PC8446-100MG
PC8446-1G
APOSPC8446-100MGEA
72.5
EUR
APOSPC8446-100MG
APOSPC8446-1G
2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile 97%
2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile
Formula:
C₁₃H₇FN₂S MW: 242,28 g/mol Melting Pt: 64…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00231908 CAS Number: 306976-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...