2-Fluoronicotinic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoropyridine-3-carboxylic acid, 2-Fluoro-3-pyridinecarboxylic acid

PC8476-100G PC8476-25G PC8476-5G
APOSPC8476-100GEA 183 EUR
APOSPC8476-100G APOSPC8476-25G APOSPC8476-5G
2-Fluoronicotinic acid 97%
2-Fluoronicotinic acid
Formula: C₆H₄FNO₂
MW: 141,1 g/mol
Melting Pt: 161…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00040744
CAS Number: 393-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR