2-[3-(Trifluoromethyl)phenoxy]ethanethioamide

Supplier: Apollo Scientific

PC8500-10G PC8500-1G PC8500-5G
APOSPC8500-10GEA 477 EUR
APOSPC8500-10G APOSPC8500-1G APOSPC8500-5G
2-[3-(Trifluoromethyl)phenoxy]ethanethioamide
2-[3-(Trifluoromethyl)phenoxy]ethanethioamide
Formula: C₉H₈F₃NOS
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00172129
CAS Number: 20293-32-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR