2-Methyl-6-(trifluoromethyl)nicotinic acid

Supplier: Apollo Scientific
Warning

PC8524-10G PC8524-5G PC8524-1G
APOSPC8524-10GEA 270 EUR
APOSPC8524-10G APOSPC8524-5G APOSPC8524-1G
2-Methyl-6-(trifluoromethyl)nicotinic acid
2-Methyl-6-(trifluoromethyl)nicotinic acid
Formula: C₈H₆F₃NO₂
MW: 205,14 g/mol
Melting Pt: 137…142 °C
Storage Temperature: Ambient
MDL Number: MFCD01311987
CAS Number: 261635-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR