(S)-2-(Trifluoromethyl)pyrrolidine 97%
Supplier: Apollo Scientific
|
Danger
|
PC8535-1G
PC8535-25G
PC8535-5G
APOSPC8535-1GEA
30.6
EUR
APOSPC8535-1G
APOSPC8535-25G
APOSPC8535-5G
(S)-2-(Trifluoromethyl)pyrrolidine 97%
(S)-2-(Trifluoromethyl)pyrrolidine
Formula:
C₅H₈F₃N MW: 139,12 g/mol Boiling Pt: 110…112 °C Melting Pt: 27…33 °C Flash Pt: 26 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03093766 CAS Number: 119580-41-5 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...