N'-Hydroxy-2-{[3-(trifluoromethyl)pyridin-2-yl]thio}ethanimidamide
Supplier: Apollo Scientific
Synonyms:
2-[3-(Trifluoromethyl)pyridin-2-ylthio]acetamide oxime
PC8538-1G
PC8538-5G
APOSPC8538-1GEA
176
EUR
APOSPC8538-1G
APOSPC8538-5G
N'-Hydroxy-2-{[3-(trifluoromethyl)pyridin-2-yl]thio}ethanimidamide
N'-Hydroxy-2-{[3-(trifluoromethyl)pyridin-2-yl]thio}ethanimidamide
Formula:
C₈H₈F₃N₃OS MW: 251,23 g/mol Melting Pt: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00208090 CAS Number: 263762-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...