1,2-Dichlorodecafluorocyclohexane

Supplier: Apollo Scientific
Warning

PC8574-5G PC8574-1G
APOSPC8574-5GEA 272 EUR
APOSPC8574-5G APOSPC8574-1G
1,2-Dichlorodecafluorocyclohexane
1,2-Dichlorodecafluorocyclohexane
Formula: C₆Cl₂F₁₀
Boiling Pt: 109…110 °C/756mmHg
Melting Pt: 32…34 °C
Storage Temperature: Ambient
MDL Number: MFCD01862047
CAS Number: 336-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR