1H,1H,9H-Perfluorononyl-p-toluenesulphonate 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1H,1H,9H-Hexadecafluoronon-1-yl toluene-4-sulphonate

PC8634-100G PC8634-25G PC8634-5G
APOSPC8634-100GEA 446 EUR
APOSPC8634-100G APOSPC8634-25G APOSPC8634-5G
1H,1H,9H-Perfluorononyl-p-toluenesulphonate 95%
1H,1H,9H-Perfluorononyl-p-toluenesulphonate
Formula: C₁₆H₁₀F₁₆O₃S
MW: 586,29 g/mol
Boiling Pt: 171,5 °C/2.3mmHg
Melting Pt: 46,5 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00085895
CAS Number: 864-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR