4-Fluorothiobenzamide

Supplier: Apollo Scientific
Danger

Synonyms: 4-Fluorobenzothioamide

PC8678-1G PC8678-5G
APOSPC8678-1GEA 102 EUR
APOSPC8678-1G APOSPC8678-5G
4-Fluorothiobenzamide
4-Fluorothiobenzamide
Formula: C₇H₆FNS
MW: 155.2 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD01313285
CAS Number: 22179-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR