5-Amino-2-fluorophenol 95+%

Supplier: Apollo Scientific
Warning

PC8698-1G PC8698-5G
APOSPC8698-1GEA 166 EUR
APOSPC8698-1G APOSPC8698-5G
5-Amino-2-fluorophenol 95+%
5-Amino-2-fluorophenol
Formula: C₆H₆FNO
MW: 127,118 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD13185335
CAS Number: 100367-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR