2-{5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid 95+%

Supplier: Apollo Scientific
Warning

PC8743-1G PC8743-5G
APOSPC8743-1GEA 336 EUR
APOSPC8743-1G APOSPC8743-5G
2-{5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid 95+%
2-{5-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Formula: C₁₃H₁₀F₃NO₂S
MW: 301,29 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08443971
CAS Number: 924868-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR